Learn More
Ethyl bromofluoroacetate, 97%
CAS: 401-55-8 | C4H6BrFO2 | 184.992 g/mol
Supplier: Thermo Scientific Chemicals B2157906
Description
Ethyl bromofluoroacetate is used as an intermediate useful in the synthesis of polyfluoroalkyl-substituted fluoro enol ethers and N-protected α-fluoroglycines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthyl bromofluoroacetate is used as an intermediate useful in the synthesis of polyfluoroalkyl-substituted fluoro enol ethers and N-protected α-fluoroglycines.
Solubility
It is soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
Specifications
| Ethyl bromofluoroacetate | |
| 1.54 | |
| 70°C (158°F) | |
| 1.4255 | |
| 5 g | |
| ethyl bromofluoroacetate, bromofluoroacetic acid ethyl ester, ethyl-2-bromofluoroacetate, ethylbromofluoroacetate, acetic acid, bromofluoro-, ethyl ester, bromofluoroacetate, ethyl fluorobromoacetate, acmc-209jb9, ksc236i3t | |
| ULNDTPIRBQGESN-UHFFFAOYSA-N | |
| ethyl 2-bromo-2-fluoroacetate | |
| 2733407 | |
| 97% |
| 401-55-8 | |
| 154°C | |
| C4H6BrFO2 | |
| MFCD00042095 | |
| UN2810 | |
| It is soluble in water. | |
| CCOC(=O)C(F)Br | |
| 184.992 | |
| 184.99 |
Chemical Identifiers
| 401-55-8 | |
| 184.992 | |
| ULNDTPIRBQGESN-UHFFFAOYSA-N | |
| 2733407 | |
| CCOC(=O)C(F)Br |
| C4H6BrFO2 | |
| MFCD00042095 | |
| ethyl bromofluoroacetate, bromofluoroacetic acid ethyl ester, ethyl-2-bromofluoroacetate, ethylbromofluoroacetate, acetic acid, bromofluoro-, ethyl ester, bromofluoroacetate, ethyl fluorobromoacetate, acmc-209jb9, ksc236i3t | |
| ethyl 2-bromo-2-fluoroacetate |
Safety and Handling
GHS H Statement
H301-H311-H331-H318-H227-H315-H335
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes serious eye damage.
Combustible liquid.
Causes skin irritation.
May cause respiratory irritation.
P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
H227-H302+H312+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber : 206-928-4
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only