missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl bromodifluoroacetate, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC305800010

 View more versions of this product

Catalog No. AC305800010

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥97.5%
Liquid
Clear colorless to slightly yellow
Authentic
1.3860 to 1.3890 (20°C, 589nm)
667-27-6
BrCF2CO2C2H5
MFCD00042069
202.99
1g
1.5830g/mL
Glass bottle
Ethyl bromodifluoroacetate
C4H5BrF2O2
98%
98%

Chemical Identifiers

667-27-6
C4H5BrF2O2
202.99
Ethyl bromodifluoroacetate, 98%
98%
BrCF2CO2C2H5
MFCD00042069
≥97.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 211-567-0