missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl benzoylformate, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 410350250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Ethyl benzoylformate | |
1.1220g/mL | |
113°C | |
94% min. (GC) | |
C10H10O3 | |
1.5150 to 1.517 | |
25g | |
ethyl benzoylformate, ethyl phenylglyoxylate, ethyl oxophenylacetate, ethylbenzoylformate, phenylglyoxylic acid ethyl ester, ethyl oxo phenyl acetate, benzeneacetic acid, .alpha.-oxo-, ethyl ester, benzoylformic acid ethyl ester, ethyl benzoyl formate, glyoxylic acid, phenyl-, ethyl ester | |
CCOC(=O)C(=O)C1=CC=CC=C1 | |
178.19 | |
CHEBI:84260 | |
95% |
1603-79-8 | |
138°C to 139°C (18.0mmHg) | |
Authentic | |
Glass bottle | |
C6H5COCO2CH2CH3 | |
MFCD00009120 | |
1.122 | |
QKLCQKPAECHXCQ-UHFFFAOYSA-N | |
ethyl 2-oxo-2-phenylacetate | |
15349 | |
178.19 |
Chemical Identifiers
1603-79-8 | |
178.19 | |
QKLCQKPAECHXCQ-UHFFFAOYSA-N | |
15349 | |
ethyl 2-oxo-2-phenylacetate |
C10H10O3 | |
MFCD00009120 | |
ethyl benzoylformate, ethyl phenylglyoxylate, ethyl oxophenylacetate, ethylbenzoylformate, phenylglyoxylic acid ethyl ester, ethyl oxo phenyl acetate, benzeneacetic acid, .alpha.-oxo-, ethyl ester, benzoylformic acid ethyl ester, ethyl benzoyl formate, glyoxylic acid, phenyl-, ethyl ester | |
CHEBI:84260 | |
CCOC(=O)C(=O)C1=CC=CC=C1 |
Safety and Handling
EINECSNumber : 216-504-
RUO â Research Use Only