Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl benzoate, 99%

$30.58 - $145.85

Chemical Identifiers

Chemical Name or Material Ethyl benzoate
CAS 93-89-0
Molecular Formula C9H10O2
Formula Weight 150.18
MDL Number MFCD00009109
Beilstein 1908172
Merck Index 14,3766
Synonym Benzoic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA130570E

  SDS
alfa aesar™
A130570E
2500g Each for $145.85

AAA1305736

  SDS
alfa aesar™
A1305736
500g Each for $43.55

AAA1305722

  SDS
alfa aesar™
A1305722
100g Each for $30.58
Description & Specifications

Specifications

Formula Weight 150.18
Melting Point −34°C
Health Hazard 3 GHS P Statement
P210-P280-P305+P351+P338-P362-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 212° to 214°C
Beilstein 1908172
Health Hazard 2 GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Assay 99%
CAS 93-89-0
Molecular Formula C9H10O2
MDL Number MFCD00009109
Synonym Benzoic acid ethyl ester
Health Hazard 1 Warning
Flash Point 84°C (183°F)
Merck Index 14,3766
TSCA TSCA
Chemical Name or Material Ethyl benzoate
Quantity 2500g
RTECS Number DH0200000

Applications
Ethyl benzoate is used as a perfume scent. It acts as a food flavoring agent. It is an active component of artificial fruit flavors. Further, it is used in cosmetics and personal care products as fragrance ingredients and preservatives.

Solubility
Miscible with methanol and diethyl ether. Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.

Safety and Handling

Warning

GHS P Statement
P210-P280-P305+P351+P338-P362-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.