Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl benzimidazole-4-carboxylate, 95%

Chemical Identifiers

Chemical Name or Material Ethyl benzimidazole-4-carboxylate
CAS 167487-83-4
Formula Weight 190.2
MDL Number MFCD18251681
Beilstein 9390984
Synonym Benzimidazole-4-carboxylic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6666903

 
alfa aesar™
H6666903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66669MD

 
alfa aesar™
H66669MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 190.2
Synonym Benzimidazole-4-carboxylic acid ethyl ester
Physical Form Solid
Assay 95%
Quantity 1g
MDL Number MFCD18251681
CAS 167487-83-4
Beilstein 9390984
Chemical Name or Material Ethyl benzimidazole-4-carboxylate