missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl alpha-Iodophenylacetate, TCI America™

Manufacturer:  TCI America E120025G

 View more versions of this product

Catalog No. E120025G



Specifications

Specifications

Ethyl alpha-Iodophenylacetate
25g
C10H11IO2
alpha-Iodophenylacetic Acid Ethyl Ester, Ethyl 2-Iodo-2-phenylacetate, 2-Iodo-2-phenylacetic Acid Ethyl Ester
CCOC(=O)C(C1=CC=CC=C1)I
290.1
290.10
≥98.0% (HPLC)
Undesignated
78489-65-3
3265
GFYUBPKXVMWINH-UHFFFAOYSA-N
ethyl 2-iodo-2-phenylacetate
13346227
Clear Liquid at 20°C

Chemical Identifiers

78489-65-3
290.1
alpha-Iodophenylacetic Acid Ethyl Ester, Ethyl 2-Iodo-2-phenylacetate, 2-Iodo-2-phenylacetic Acid Ethyl Ester
ethyl 2-iodo-2-phenylacetate
C10H11IO2
GFYUBPKXVMWINH-UHFFFAOYSA-N
13346227
CCOC(=O)C(C1=CC=CC=C1)I
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Freezer

Documents