Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl alpha,alpha-difluorophenylacetate, 99%

$272.45 - $1,028.24

Chemical Identifiers

Chemical Name or Material Ethyl alpha,alpha-difluorophenylacetate
CAS 2248-46-6
Molecular Formula C10H10F2O2
Formula Weight 200.19
MDL Number MFCD08275296
Synonym alpha,alpha-Difluorophenylacetic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2602903

  SDS
alfa aesar™
H2602903
1g Each for $272.45

AAH2602906

  SDS
alfa aesar™
H2602906
5g Each for $1,028.24
Description & Specifications

Specifications

Formula Weight 200.19
Chemical Name or Material Ethyl alpha,alpha-difluorophenylacetate
Assay 99%
Molecular Formula C10H10F2O2
MDL Number MFCD08275296
CAS 2248-46-6
Synonym alpha,alpha-Difluorophenylacetic acid ethyl ester
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.