missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl Alcohol Standard, 10% (v/v), Ricca Chemical
$359.81 - $359.81
Chemical Identifiers
CAS | 64-17-5 |
---|---|
Molecular Formula | C2H6O |
InChI Key | LFQSCWFLJHTTHZ-UHFFFAOYSA-N |
IUPAC Name | ethanol |
SMILES | CCO |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
R2919100500
|
Ricca Chemical Company
R2919100500A |
500 mL | Natural Poly Bottle |
Each for $359.81
|
|
||||
Chemical Identifiers
64-17-5 | |
LFQSCWFLJHTTHZ-UHFFFAOYSA-N | |
CCO |
C2H6O | |
ethanol |
Specifications
64-17-5,64-17-5 | |
500 mL | |
91.0,11.0 | |
LFQSCWFLJHTTHZ-UHFFFAOYSA-N | |
ethanol | |
10% (v/v) |
Natural Poly Bottle | |
89.0,9.0 | |
C2H6O | |
CCO | |
46.07 | |
Ethyl Alcohol Standard |