Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl adamantane-1-carboxylate, 98+%

$74.51 - $1,009.77

Chemical Identifiers

Chemical Name or Material Ethyl adamantane-1-carboxylate
CAS 2094-73-7
Molecular Formula C13H20O2
Formula Weight 208.3
MDL Number MFCD00078272
Beilstein 2050027
Synonym Adamantane-1-carboxylic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2451804

 
alfa aesar™
B2451804
2g Each for $74.51

AAB2451809

 
alfa aesar™
B2451809
10g Each for $243.20

AAB2451818

 
alfa aesar™
B2451818
50g Each for $1,009.77
Description & Specifications

Specifications

Formula Weight 208.3
Chemical Name or Material Ethyl adamantane-1-carboxylate
Assay ≥98%
Molecular Formula C13H20O2
MDL Number MFCD00078272
Beilstein 2050027
Synonym Adamantane-1-carboxylic acid ethyl ester
Boiling Point 90° to 91°C (1mmHg)
Quantity 2g
CAS 2094-73-7