missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl acrylate, 99%, stab. with ca 20ppm 4-methoxyphenol

Manufacturer:  Alfa Aesar™ A154930F

 View more versions of this product

Catalog No. AAA154930F



Description

Description

Applications
Ethyl acrylate is used in the syntheses of various pharmaceutical intermediates, textiles and non-woven fibers. It is widely used in the production of polymers including resins, plastics, rubber and denture material. It is actively involved in the preparation of dimethylaminoethylacrylate (DMAEA) which is used as a monomer to make flocculants for water treatment. It plays a vital role in the manufacture of water-based latex paints and adhesives, textile and paper coatings and leather finish resins.

Solubility
Miscible with ethanol, ethyl ether and chloroform. Silghtly miscible with water and dimethyl sulfoxide.

Notes
Store in cool place. Incompatible with strong oxidizing agents, strong acids, amines, peroxides, chlorosulfonic acid, moisture and strong alkalies.
Specifications

Specifications

Ethyl acrylate
Pungent
-71°C
stabilized with ≈20ppm 4-methoxyphenol
C5H8O2
MFCD00009188
773866
ethyl acrylate, acrylic acid ethyl ester, ethyl propenoate, 2-propenoic acid, ethyl ester, ethyl 2-propenoate, ethylacrylaat, ethylakrylat, etil acrilato, acrylic acid, ethyl ester, aethylacrylat
CCOC(=O)C=C
100.117
CHEBI:82327
99%
−1°C (30°F)
99°C to 100°C
2500mL
140-88-5
CH2=CHCO2CH2CH3
UN1917
14,3759
JIGUQPWFLRLWPJ-UHFFFAOYSA-N
ethyl prop-2-enoate
8821
100.12

Chemical Identifiers

140-88-5
100.117
JIGUQPWFLRLWPJ-UHFFFAOYSA-N
8821
ethyl prop-2-enoate
C5H8O2
MFCD00009188
ethyl acrylate, acrylic acid ethyl ester, ethyl propenoate, 2-propenoic acid, ethyl ester, ethyl 2-propenoate, ethylacrylaat, ethylakrylat, etil acrilato, acrylic acid, ethyl ester, aethylacrylat
CHEBI:82327
CCOC(=O)C=C
Safety and Handling

Safety and Handling

GHS H Statement
H225-H302-H312-H332-H315-H319-H317-H335
Highly flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHYL ACRYLATE, STABILIZED

RTECSNumber : AT0700000

TSCA : TSCA

Recommended Storage : 2° to 8°C

Documents