Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl acetopyruvate, 98%

$68.11 - $824.20

Chemical Identifiers

Chemical Name or Material Ethyl acetopyruvate
CAS 615-79-2
Molecular Formula C7H10O4
Formula Weight 158.15
MDL Number MFCD00009124
Beilstein 607062
Synonym Acetopyruvic acid ethyl ester; Ethyl 2,4-dioxovalerate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1380806

  SDS
alfa aesar™
A1380806
5g Each for $68.11

AAA1380814

  SDS
alfa aesar™
A1380814
25g Each for $230.54

AAA1380822

  SDS
alfa aesar™
A1380822
100g Each for $824.20
Description & Specifications

Specifications

Formula Weight 158.15
Chemical Name or Material Ethyl acetopyruvate
Assay 98%
Molecular Formula C7H10O4
MDL Number MFCD00009124
CAS 615-79-2
Beilstein 607062
Synonym Acetopyruvic acid ethyl ester; Ethyl 2,4-dioxovalerate
Recommended Storage 2° to 8°C
Flash Point >110°C (230°F)
Boiling Point 108° to 109°C (12mmHg)
Melting Point 16° to 18°C
Quantity 5g

Applications
Ethyl acetopyruvate is used in the preparation of 5-methyl-isoxazole-3-carboxylic acid ethyl ester. It is involved in the preparation of ethyl 5-amino-2-methylpyndine-4-carboxyIate by reacting with nitroacetamide.

Solubility
Miscible with water, ether,chloroform, methanol and ethanol.

Notes
Hygroscopic. Store in a cool place. Incompatible with acids, bases, oxidizing agents and reducing agents.

Safety and Handling

2° to 8°C