Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl acetoacetate propylene glycol ketal, 97%

$192.86 - $192.86

Chemical Identifiers

Chemical Name or Material Ethyl acetoacetate propylene glycol ketal
CAS 6290-17-1
Molecular Formula C9H16O4
Formula Weight 188.23
MDL Number MFCD00151819
Beilstein 138927
Synonym 2,4-Dimethyl-1,3-dioxolane-2-acetic acid ethyl ester; Ethyl 2,4-dimethyl-1,3-dioxolane-2-acetate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1351422

  SDS
alfa aesar™
L1351422
100g Each for $192.86
Description & Specifications

Specifications

Formula Weight 188.23
Molecular Formula C9H16O4
Chemical Name or Material Ethyl acetoacetate propylene glycol ketal
Assay 97%
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H227
Combustible liquid.
CAS 6290-17-1
Health Hazard 3 GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00151819
TSCA TSCA
Flash Point 65°C (149°F)
Synonym 2,4-Dimethyl-1,3-dioxolane-2-acetic acid ethyl ester; Ethyl 2,4-dimethyl-1,3-dioxolane-2-acetate
Quantity 100g
Boiling Point 90°C (10mmHg)
Beilstein 138927
Safety and Handling

Warning

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H227
Combustible liquid.