missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl acetoacetate propylene glycol ketal, 97%

Manufacturer:  Alfa Aesar™ L1351422

 View more versions of this product

Catalog No. AAL1351422



Specifications

Specifications

Ethyl acetoacetate propylene glycol ketal
90°C (10mmHg)
6290-17-1
MFCD00151819
ethyl 2-2,4-dimethyl-1,3-dioxolan-2-yl acetate, 1,3-dioxolane-2-acetic acid, 2,4-dimethyl-, ethyl ester, ethyl acetoacetate propylene glycol ketal, fraistone, ethyl 2,4-dimethyl-1,3-dioxolane-2-acetate, ethyl dimethyl dioxolane acetate, acetoacetic acid, ethyl ester, 1,2-propylene ketal, cis-and trans-ethyl 2,4-dimethyl-1,3-dioxolane-2-acetate, ethyl 2,4-dimethyl-1,3-dioxolan-2-yl acetate, 2,4-dimethyl-1,3-dioxolane-2-acetic acid ethyl ester
CCOC(=O)CC1(OCC(O1)C)C
188.223
188.23
65°C (149°F)
100g
C9H16O4
138927
GSIXJEIRJVOUFB-UHFFFAOYSA-N
ethyl 2-(2,4-dimethyl-1,3-dioxolan-2-yl)acetate
95392
97%

Chemical Identifiers

6290-17-1
188.223
GSIXJEIRJVOUFB-UHFFFAOYSA-N
95392
CCOC(=O)CC1(OCC(O1)C)C
C9H16O4
MFCD00151819
ethyl 2-2,4-dimethyl-1,3-dioxolan-2-yl acetate, 1,3-dioxolane-2-acetic acid, 2,4-dimethyl-, ethyl ester, ethyl acetoacetate propylene glycol ketal, fraistone, ethyl 2,4-dimethyl-1,3-dioxolane-2-acetate, ethyl dimethyl dioxolane acetate, acetoacetic acid, ethyl ester, 1,2-propylene ketal, cis-and trans-ethyl 2,4-dimethyl-1,3-dioxolane-2-acetate, ethyl 2,4-dimethyl-1,3-dioxolan-2-yl acetate, 2,4-dimethyl-1,3-dioxolane-2-acetic acid ethyl ester
ethyl 2-(2,4-dimethyl-1,3-dioxolan-2-yl)acetate
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Documents