missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl acetoacetate, 99+%, extra pure, Thermo Scientific Chemicals
$50.64 - $159.15
Chemical Identifiers
CAS | 141-97-9 |
---|---|
Molecular Formula | C6H10O3 |
Molecular Weight (g/mol) | 130.14 |
MDL Number | MFCD00009199 |
InChI Key | XYIBRDXRRQCHLP-UHFFFAOYSA-N |
Synonym | ethyl acetoacetate, ethyl acetylacetate, ethyl 3-oxobutyrate, diacetic ether, butanoic acid, 3-oxo-, ethyl ester, ethyl acetylacetonate, acetoacetic acid, ethyl ester, 3-oxobutanoic acid ethyl ester, 3-oxo-butyric acid ethyl ester, active acetylacetate |
PubChem CID | 8868 |
ChEBI | CHEBI:4893 |
IUPAC Name | ethyl 3-oxobutanoate |
SMILES | CCOC(=O)CC(=O)C |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC220401000
|
Thermo Scientific Chemicals
220401000 |
100 g | Glass bottle |
Each for $50.64
|
|
||||
AC220405000
|
Thermo Scientific Chemicals
220405000 |
500 g | Glass bottle |
Each for $62.62
|
|
||||
AC220400025
|
Thermo Scientific Chemicals
220400025 |
2.5 kg | Glass bottle |
Each for $159.15
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
141-97-9 | |
130.14 | |
XYIBRDXRRQCHLP-UHFFFAOYSA-N | |
8868 | |
ethyl 3-oxobutanoate |
C6H10O3 | |
MFCD00009199 | |
ethyl acetoacetate, ethyl acetylacetate, ethyl 3-oxobutyrate, diacetic ether, butanoic acid, 3-oxo-, ethyl ester, ethyl acetylacetonate, acetoacetic acid, ethyl ester, 3-oxobutanoic acid ethyl ester, 3-oxo-butyric acid ethyl ester, active acetylacetate | |
CHEBI:4893 | |
CCOC(=O)CC(=O)C |
Specifications
141-97-9 | |
1.0300g/mL | |
70°C | |
99% min. (GC) | |
C6H10O3 | |
CH3COCH2CO2C2H5 | |
100 g | |
12,222 | |
15, 3812 | |
Solubility in water: 116g/L (20°C). Other solubilities: soluble in most organic solvent: diethyl ether,,benzene,ethanol,ehyl acetate,chloroform,,acetone | |
CCOC(=O)CC(=O)C | |
130.14 | |
1.696 mPa.s (20°C) | |
130.14 | |
Extra Pure | |
Ethyl acetoacetate, p.a. |
-45°C | |
180°C | |
Authentic | |
Glass bottle | |
1.4180 to 1.42 | |
MFCD00009199 | |
03, 632 | |
1.03 | |
ethyl acetoacetate, ethyl acetylacetate, ethyl 3-oxobutyrate, diacetic ether, butanoic acid, 3-oxo-, ethyl ester, ethyl acetylacetonate, acetoacetic acid, ethyl ester, 3-oxobutanoic acid ethyl ester, 3-oxo-butyric acid ethyl ester, active acetylacetate | |
XYIBRDXRRQCHLP-UHFFFAOYSA-N | |
ethyl 3-oxobutanoate | |
8868 | |
CHEBI:4893 | |
99+% | |
Liquid |
Safety and Handling
EINECSNumber : 205-516-1
RTECSNumber : AK5250000
TSCA : TSCA
RUO – Research Use Only