missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl acetoacetate, 99+%, extra pure, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 220400025

 View more versions of this product

Catalog No. AC220400025



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ethyl acetoacetate
141-97-9
1.0300g/mL
70°C
99% min. (GC)
C6H10O3
CH3COCH2CO2C2H5
2.5 kg
12,222
15, 3812
Solubility in water: 116g/L (20°C). Other solubilities: soluble in most organic solvent: diethyl ether,,benzene,ethanol,ehyl acetate,chloroform,,acetone
CCOC(=O)CC(=O)C
130.14
1.696 mPa.s (20°C)
130.14
Extra Pure
p.a.
-45°C
180°C
Authentic
Glass bottle
1.4180 to 1.42
MFCD00009199
03, 632
1.03
ethyl acetoacetate, ethyl acetylacetate, ethyl 3-oxobutyrate, diacetic ether, butanoic acid, 3-oxo-, ethyl ester, ethyl acetylacetonate, acetoacetic acid, ethyl ester, 3-oxobutanoic acid ethyl ester, 3-oxo-butyric acid ethyl ester, active acetylacetate
XYIBRDXRRQCHLP-UHFFFAOYSA-N
ethyl 3-oxobutanoate
8868
CHEBI:4893
99+%
Liquid

Chemical Identifiers

141-97-9
130.14
XYIBRDXRRQCHLP-UHFFFAOYSA-N
8868
ethyl 3-oxobutanoate
C6H10O3
MFCD00009199
ethyl acetoacetate, ethyl acetylacetate, ethyl 3-oxobutyrate, diacetic ether, butanoic acid, 3-oxo-, ethyl ester, ethyl acetylacetonate, acetoacetic acid, ethyl ester, 3-oxobutanoic acid ethyl ester, 3-oxo-butyric acid ethyl ester, active acetylacetate
CHEBI:4893
CCOC(=O)CC(=O)C
Safety and Handling

Safety and Handling

EINECSNumber : 205-516-1

RTECSNumber : AK5250000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only