missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl acetimidate hydrochloride, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 224102500

 View more versions of this product

Catalog No. AC224102500

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥96.0%
123.58
White
112680
Authentic
97%
Ethyl acetimidate hydrochloride
C4H9NO·HCl
Crystalline Powder
250g
02, 182
Glass bottle
97%
MFCD00012572
CH3C(=NH)OC2H5·HCl

Chemical Identifiers

112680
C4H9NO·HCl
123.58
Ethyl acetimidate hydrochloride, 97%
97%
CH3C(=NH)OC2H5·HCl
MFCD00012572
≥96.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

MOISTURE SENSITIVE

EINECSNumber : 218-631-7

TSCA : TSCA