Please login to your online account to display your discounted pricing

Ethyl acetimidate hydrochloride, 97%, ACROS Organics™


Quantity 50g
Packaging Glass bottle

Manufacturer: fisher scientific acros  224100500

 View more versions of this product

Catalog No. AC224100500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Assay ≥96.0%
Formula Weight 123.58
Quantity 50g
Color White
EINECS Number 218-631-7
TSCA TSCA
Beilstein 02, 182
CAS 112680
Chemical Name or Material Ethyl acetimidate hydrochloride
Packaging Glass bottle
Name Note 97%
Molecular Formula C4H9NO·HCl
Linear Formula CH3C(=NH)OC2H5·HCl
Health Hazard 1 MOISTURE SENSITIVE
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Assay Percent Range 97%
MDL Number MFCD00012572

Specifications

Chemical Name or Material Ethyl acetimidate hydrochloride
Name Note 97%
CAS 112680
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C4H9NO·HCl
Linear Formula CH3C(=NH)OC2H5·HCl
Formula Weight 123.58
MDL Number MFCD00012572
Accessories

MOISTURE SENSITIVE

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents