Please login to your online account to display your discounted pricing

Ethyl acetimidate hydrochloride, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Ethyl acetimidate hydrochloride
Name Note 97%
CAS 112680
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C4H9NO·HCl
Linear Formula CH3C(=NH)OC2H5·HCl
Formula Weight 123.58
MDL Number MFCD00012572
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC224100500

  SDS
fisher scientific acros
224100500
Glass bottle 50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC224102500

  SDS
thermo fisher scientific
224102500
Glass bottle 250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Assay ≥96.0%
Formula Weight 123.58
Quantity 50g
Color White
EINECS Number 218-631-7
TSCA TSCA
Beilstein 02, 182
CAS 112680
Chemical Name or Material Ethyl acetimidate hydrochloride
Packaging Glass bottle
Name Note 97%
Molecular Formula C4H9NO·HCl
Linear Formula CH3C(=NH)OC2H5·HCl
Health Hazard 1 MOISTURE SENSITIVE
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Assay Percent Range 97%
MDL Number MFCD00012572
Safety and Handling

MOISTURE SENSITIVE

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.