missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl acetimidate hydrochloride, 97%

$87.43 - $341.72

Chemical Identifiers

CAS 2208-07-3
Molecular Formula C4H9NO·HCl
Formula Weight 123.58
MDL Number MFCD00012572
Beilstein 3552401
Synonym Ethyl iminoacetate hydrochloride
Chemical Name or Material Ethyl acetimidate hydrochloride
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1861818 SDS Alfa Aesar™
A1861818
50g Each for $87.43
AAA1861830 SDS Alfa Aesar™
A1861830
250g Each for $341.72
AAA1861809 SDS Alfa Aesar™
A1861809
10g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

Applications
Ethyl acetimidate hydrochloride was used in preparation of amidinated carbonic anhydrase via chemical modification of human erythrocyte carbonic anhydrase. It was also used in synthesis of methyl 2-methyl-2-thiazoline-4-carboxylate hydrochloride.

Solubility
Soluble in water (50mg/L).

Notes
Moisture Sensitive, store away from water/moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under dry inert gas.
Description & Specifications

Chemical Identifiers

2208-07-3
123.58
3552401
Ethyl acetimidate hydrochloride
C4H9NO·HCl
MFCD00012572
Ethyl iminoacetate hydrochloride

Specifications

2208-07-3
123.58
3552401
≈108°C (dec.)
Ethyl acetimidate hydrochloride
C4H9NO·HCl
MFCD00012572
Ethyl iminoacetate hydrochloride
50g
97%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA