Please login to your online account to display your discounted pricing

Ethyl Acetate (Sequencing), Fisher BioReagents


Quantity 4L
Packaging Glass Bottle

Manufacturer: fisher scientific company  11254

 View more versions of this product

Catalog No. BP1125-4

  • / Each
  • / Case of 4 Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.9%
Formula Weight 88.11
Boiling Point 75°C
Purity Grade Sequencing Grade
Melting Point −83.5°C
Quantity 4L
Merck Index 15, 3811
DOT Information DOT Class 3, : Flammable Liquid
Physical Form Liquid
Color Colorless
Density 0.902g/cm³
Viscosity 0.45 cP at 20°C
Vapor Pressure 103mbar at 20°C
Absorbance 0.025 max. at 270nm
Peroxides Not Detected Detected/Not Detect.
Packaging Glass Bottle
Assay Percent Range ≥99.9 %
Refractive Index 1.3680 to 1.3710 (at 25°C)
Health Hazard 2 DANGER!
Health Hazard 3 Emergency Overview
Highly flammable. Vapors may cause drowsiness and dizziness. Repeated exposure may cause skin dryness or cracking. May cause central nervous system effects. May cause skin and respiratory tract irritation. May be harmful by inhalation, in contact with skin and if swallowed. Use only under a chemical fume hood. Use explosion-proof equipment. Wear personal protective equipment. Do not breathe vapors or spray mist. Do not get in eyes, on skin, or on clothing. Keep away from open flames, hot surfaces and sources of ignition. Keep away from open flames, hot surfaces and sources of ignition. Use only non-sparking tools.

NFPA

Health:1
Flammability:3
Instability:0
Chemical Name or Material Ethyl Acetate
Linear Formula CH3COOCH2CH3
ChemAlert Storage Symbol Red
UN Number 1173
Residue after Evaporation 10ppm max.
Substances Darkened by H2SO4 Pass Test
Additional Information Vapor Pressure: 103mbar at 20°C
Water 0.02% max.
Fluorescence Background Pass Test
Titratable Acid 0.0009mEq/g max.
CAS 141-78-6
Recommended Storage RT
Cleanup Symbol Solvent
Molecular Formula C4H8O2

Specifications

Chemical Name or Material Ethyl Acetate
CAS 141-78-6
Purity Grade Sequencing Grade
Assay ≥99.9%
Assay Percent Range ≥99.9 %
Molecular Formula C4H8O2
Linear Formula CH3COOCH2CH3
Formula Weight 88.11
Accessories

Accessories for Catalog Number: BP11254

Item Mfr. No. Item Description Price Per Unit Quantity  
03-439
Eagle™ Thermoplastics™ Safety Bottle Carriers
fisher scientific
B102
Safety bottle carriers are molded-in handle Each for $217.50
Accessories
 

Emergency Overview
Highly flammable. Vapors may cause drowsiness and dizziness. Repeated exposure may cause skin dryness or cracking. May cause central nervous system effects. May cause skin and respiratory tract irritation. May be harmful by inhalation, in contact with skin and if swallowed. Use only under a chemical fume hood. Use explosion-proof equipment. Wear personal protective equipment. Do not breathe vapors or spray mist. Do not get in eyes, on skin, or on clothing. Keep away from open flames, hot surfaces and sources of ignition. Keep away from open flames, hot surfaces and sources of ignition. Use only non-sparking tools.

NFPA

Health:1
Flammability:3
Instability:0

DANGER!

RT

 

SDS & Safety Documents