Please login to your online account to display your discounted pricing

Ethyl Acetate (Sequencing), Fisher BioReagents


Quantity 1L
Packaging Glass Bottle

Manufacturer: fisher scientific company  11251

 View more versions of this product

Catalog No. BP1125-1

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material Ethyl Acetate
Cleanup Symbol Solvent
Absorbance 0.025 max. at 270nm
CAS 141-78-6
ChemAlert Storage Symbol Red
Health Hazard 2 DANGER!
Health Hazard 3 Emergency Overview
Highly flammable. Vapors may cause drowsiness and dizziness. Repeated exposure may cause skin dryness or cracking. May cause central nervous system effects. May cause skin and respiratory tract irritation. May be harmful by inhalation, in contact with skin and if swallowed. Use only under a chemical fume hood. Use explosion-proof equipment. Wear personal protective equipment. Do not breathe vapors or spray mist. Do not get in eyes, on skin, or on clothing. Keep away from open flames, hot surfaces and sources of ignition. Keep away from open flames, hot surfaces and sources of ignition. Use only non-sparking tools.

NFPA

Health:1
Flammability:3
Instability:0
Recommended Storage RT
DOT Information DOT Class 3, : Flammable Liquid
Additional Information Vapor Pressure: 103mbar at 20°C
Purity Grade Sequencing Grade
Physical Form Liquid
Assay ≥99.9%
Assay Percent Range ≥99.9 %
Molecular Formula C4H8O2
Linear Formula CH3COOCH2CH3
Formula Weight 88.11
UN Number 1173
Merck Index 15, 3811
Density 0.902g/cm³
Fluorescence Background Pass Test
Packaging Glass Bottle
Peroxides Not Detected Detected/Not Detect.
Refractive Index 1.3680 to 1.3710 (at 25°C)
Residue after Evaporation 10ppm max.
Substances Darkened by H2SO4 Pass Test
Titratable Acid 0.0009mEq/g max.
Vapor Pressure 103mbar at 20°C
Water 0.02% max.
Vapor Density 3.04g/mol
Viscosity 0.45 cP at 20°C
Boiling Point 75°C
Color Colorless
Melting Point −83.5°C
Quantity 1L

Specifications

Chemical Name or Material Ethyl Acetate
CAS 141-78-6
Purity Grade Sequencing Grade
Assay ≥99.9%
Assay Percent Range ≥99.9 %
Molecular Formula C4H8O2
Linear Formula CH3COOCH2CH3
Formula Weight 88.11
Accessories
 

Emergency Overview
Highly flammable. Vapors may cause drowsiness and dizziness. Repeated exposure may cause skin dryness or cracking. May cause central nervous system effects. May cause skin and respiratory tract irritation. May be harmful by inhalation, in contact with skin and if swallowed. Use only under a chemical fume hood. Use explosion-proof equipment. Wear personal protective equipment. Do not breathe vapors or spray mist. Do not get in eyes, on skin, or on clothing. Keep away from open flames, hot surfaces and sources of ignition. Keep away from open flames, hot surfaces and sources of ignition. Use only non-sparking tools.

NFPA

Health:1
Flammability:3
Instability:0

DANGER!

RT

 

SDS & Safety Documents