missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl Acetate, Reagent, ACS, 99.5%, Spectrum™

E1025, 141-78-6, C4H8O2

$254.54 - $6,206.66

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
IUPAC Name ethyl acetate
SMILES CCOC(C)=O
View More Specs

Products 8
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18602503
View Documents
Spectrum Chemical Mfg Cor
E1025500MLGL
500 mL Amber Glass Bottle
Each for $254.54
 
18602504
View Documents
Spectrum Chemical Mfg Cor
E1025500MLGLCS6
6 x 500 mL Amber Glass Bottle
Case of 6 Each for $975.71
 
18602508
View Documents
Spectrum Chemical Mfg Cor
E10251LTGL
1 L Amber Glass Bottle
Each for $364.63
 
18602509
View Documents
Spectrum Chemical Mfg Cor
E10251LTGLCS6
6 x 1 L Amber Glass Bottle
Case of 6 Each for $1,690.71
 
18602505
View Documents
Spectrum Chemical Mfg Cor
E10254LTGL
4 L Amber Glass Bottle
Each for $894.03
 
18602506
View Documents
Spectrum Chemical Mfg Cor
E10254LTGLCS4
4 x 4 L Amber Glass Bottle
Case of 4 Each for $2,359.26
 
18602507
View Documents
Spectrum Chemical Mfg Cor
E102520LTMT
20 L Steel Can
Each for $2,352.14
 
18602510
View Documents
Spectrum Chemical Mfg Cor
E1025200LTBL
200 L Drum
Each for $6,206.66
 
Description

Description

Spectrum™ Ethyl Acetate, Reagent, ACS , is a colorless liquid that smells like pear drops. Abbreviated as EA, it can be found in glues, nail polish removers, cigarettes and other products. Primarily used as a diluent because of its low cost and toxicity, it is also used in laboratory settings for column chromatography and extractions. Prone to hydrolysis and transesterification, it is rarely used as a solvent. As an ACS grade Reagent, Spectrum Chemical manufactured Ethyl Acetate is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness.
Specifications

Chemical Identifiers

141-78-6
88.11
ethyl acetate
C4H8O2
XEKOWRVHYACXOJ-UHFFFAOYSA-N
CCOC(C)=O

Specifications

141-78-6
C4H8O2
XEKOWRVHYACXOJ-UHFFFAOYSA-N
ethyl acetate
99.5%
Amber Glass Bottle
0.2%
100%
UN1173
CCOC(C)=O
88.11
ACS
0.003%
500 mL
SDS