Please login to your online account to display your discounted pricing

Ethyl Acetate (NF), Fisher Chemical

$447.50 - $1,583.00

Chemical Identifiers

Chemical Name or Material Ethyl Acetate
CAS 141-78-6
Purity Grade BP/EP/FCC/JP/NF/USP
Assay 99.0 to 100.5% (NF)
Assay Percent Range 99.0 to 100.5 %
Molecular Formula C4H8O2
Linear Formula CH3COOC2H5
Formula Weight 88.11
MDL Number MFCD00009171
Synonym Acetic Acid Ethyl Ester
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

E124-20

  SDS
fisher scientific company
E12420
Steel Pail 20L Each for $1,583.00

E124-4

  Accessories SDS
fisher scientific company
E1244
Glass Bottle 4L Each for $447.50
Case of 4 Each for $1,790.00
Description & Specifications

Specifications

Purity Grade BP/EP/FCC/JP/NF/USP
Boiling Point 75°C
Melting Point −83.5°C
Formula Weight 88.11
Quantity 20L
UN Number 1173
DOT Information DOT Class 3, : Flammable Liquid
Vapor Pressure 103mbar at 20°C
Vapor Density 3.04g/mol
Viscosity 0.45 cP at 20°C
Physical Form Liquid
Color Colorless
Assay 99.0 to 100.5% (NF)
Density 0.902g/cm³
Additional Information Vapor Pressure: 103mbar at 20°C
Non_volatile Matter 0.02% max.
Packaging Steel Pail
Purity 99.5% min.
Residual Solvents Meets Requirements (USP)
MDL Number MFCD00009171
ChemAlert Storage Symbol Red
Carbonizable Substances Pass Test
Health Hazard 3 Emergency Overview
Highly flammable. Vapors may cause drowsiness and dizziness. Repeated exposure may cause skin dryness or cracking. May cause central nervous system effects. May cause skin and respiratory tract irritation. May be harmful by inhalation, in contact with skin and if swallowed. Use only under a chemical fume hood. Use explosion-proof equipment. Wear personal protective equipment. Do not breathe vapors or spray mist. Do not get in eyes, on skin, or on clothing. Keep away from open flames, hot surfaces and sources of ignition. Keep away from open flames, hot surfaces and sources of ignition. Use only non-sparking tools.

NFPA

Health:1
Flammability:3
Instability:0
Identification Pass Test
Methyl Compounds Pass Test
Specific Gravity 0.894 to 0.898 (NF)
Assay Percent Range 99.0 to 100.5 %
Synonym Acetic Acid Ethyl Ester
Health Hazard 2 DANGER!
Acidity Pass Test (NF)
Chemical Name or Material Ethyl Acetate
Cleanup Symbol Solvent
Molecular Formula C4H8O2
Linear Formula CH3COOC2H5
CAS 141-78-6

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling
 

Emergency Overview
Highly flammable. Vapors may cause drowsiness and dizziness. Repeated exposure may cause skin dryness or cracking. May cause central nervous system effects. May cause skin and respiratory tract irritation. May be harmful by inhalation, in contact with skin and if swallowed. Use only under a chemical fume hood. Use explosion-proof equipment. Wear personal protective equipment. Do not breathe vapors or spray mist. Do not get in eyes, on skin, or on clothing. Keep away from open flames, hot surfaces and sources of ignition. Keep away from open flames, hot surfaces and sources of ignition. Use only non-sparking tools.

NFPA

Health:1
Flammability:3
Instability:0

DANGER!