missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl acetate, HPLC Grade, >99.5%, Alfa Aesar™

Maximum level of impurities: Evaporation residue 3ppm, Chlorinated pesticides 5ppt, H2O 0.05%

Manufacturer:  Alfa Aesar 22912M1

 View more versions of this product

Catalog No. AA22912M1



Specifications

Specifications

Ethyl acetate
HPLC
>99.5%
MFCD00009171
XEKOWRVHYACXOJ-UHFFFAOYSA-N
ethyl acetate
8857
88.11
460°C (860°F)
Colorless
141-78-6
Colorless liquid
C4H8O2
ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
CCOC(=O)C
88.106
CHEBI:27750
Liquid
Fruit-like
4 x 1L

Chemical Identifiers

141-78-6
88.106
XEKOWRVHYACXOJ-UHFFFAOYSA-N
8857
ethyl acetate
C4H8O2
MFCD00009171
ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
CHEBI:27750
CCOC(=O)C
Safety and Handling

Safety and Handling

H225-H319-H336: Highly flammable liquid and vapour. Causes serious eye irritation. May cause drowsiness or dizziness.

DOTInformation : Hazard Class: 3, Packing Group: II

EINECSNumber : 205-500-4

SDS Documents