Please login to your online account to display your discounted pricing

Ethyl Acetate (HPLC), Fisher Chemical


Quantity 1L
Packaging Glass Bottle, Safe-Cote™

Manufacturer: fisher scientific company  E195SK1

 View more versions of this product

Catalog No. E195SK-1

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.9%
Boiling Point 75°C
Purity Grade HPLC Grade
Formula Weight 88.11
Melting Point −83.5°C
Quantity 1L
Purity Grade Notes Mobile phase for HPLC applications
DOT Information DOT Class 3, : Flammable Liquid
Vapor Pressure 103mbar at 20°C
Physical Form Liquid
Density 0.902g/cm³
Color Colorless
Viscosity 0.45 cP at 20°C
UN Number 1173
Absorbance 0.025 AU max. at 270nm
Packaging Glass Bottle, Safe-Cote™
Refractive Index 1.3680 to 1.3710 (at 25°C)
Synonym Acetic Acid Ethyl Ester
Assay Percent Range Assay: 99.9% min by GC
Substances Darkened by H2SO4 Pass Test
Health Hazard 2 DANGER!
Cleanup Symbol Solvent
ChemAlert Storage Symbol Red
MDL Number MFCD00009171
Water 0.02% max.
Additional Information Vapor Pressure: 103mbar at 20°C
Residue after Evaporation 0.001% max.
Titratable Acid 0.0009mEq/g max.
Identification Pass Test
Fluorescence Background Pass Test
Health Hazard 3 Emergency Overview
Highly flammable. Vapors may cause drowsiness and dizziness. Repeated exposure may cause skin dryness or cracking. May cause central nervous system effects. May cause skin and respiratory tract irritation. May be harmful by inhalation, in contact with skin and if swallowed. Use only under a chemical fume hood. Use explosion-proof equipment. Wear personal protective equipment. Do not breathe vapors or spray mist. Do not get in eyes, on skin, or on clothing. Keep away from open flames, hot surfaces and sources of ignition. Keep away from open flames, hot surfaces and sources of ignition. Use only non-sparking tools.

NFPA

Health:1
Flammability:3
Instability:0
Linear Formula CH3COOC2H5
Chemical Name or Material Ethyl Acetate
Molecular Formula C4H8O2
CAS 141-78-6

Specifications

Chemical Name or Material Ethyl Acetate
CAS 141-78-6
Purity Grade HPLC Grade
Purity Grade Notes Mobile phase for HPLC applications
Assay ≥99.9%
Assay Percent Range Assay: 99.9% min by GC
Molecular Formula C4H8O2
Linear Formula CH3COOC2H5
Formula Weight 88.11
MDL Number MFCD00009171
Synonym Acetic Acid Ethyl Ester
Accessories
 

Emergency Overview
Highly flammable. Vapors may cause drowsiness and dizziness. Repeated exposure may cause skin dryness or cracking. May cause central nervous system effects. May cause skin and respiratory tract irritation. May be harmful by inhalation, in contact with skin and if swallowed. Use only under a chemical fume hood. Use explosion-proof equipment. Wear personal protective equipment. Do not breathe vapors or spray mist. Do not get in eyes, on skin, or on clothing. Keep away from open flames, hot surfaces and sources of ignition. Keep away from open flames, hot surfaces and sources of ignition. Use only non-sparking tools.

NFPA

Health:1
Flammability:3
Instability:0

DANGER!

 

SDS & Safety Documents