Please login to your online account to display your discounted pricing

Ethyl Acetate (HPLC), Fisher Chemical


Quantity 19L
Packaging NOWPak™ I

Manufacturer: fisher scientific company  E195N119

 View more versions of this product

Catalog No. E195N119

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.9%
Formula Weight 88.11
Purity Grade HPLC Grade
Melting Point −83.5°C
Boiling Point 75°C
Quantity 19L
Purity Grade Notes Mobile phase for HPLC applications
DOT Information DOT Class 3, : Flammable Liquid
Physical Form Liquid
Vapor Pressure 103mbar at 20°C
Density 0.902g/cm³
Color Colorless
Viscosity 0.45 cP at 20°C
UN Number 1173
Absorbance 0.025 AU max. at 270nm
Refractive Index 1.3680 to 1.3710 (at 25°C)
Titratable Acid 0.0009mEq/g max.
Additional Information Vapor Pressure: 103mbar at 20°C
Substances Darkened by H2SO4 Pass Test
Assay Percent Range Assay: 99.9% min by GC
Health Hazard 2 DANGER!
Cleanup Symbol Solvent
ChemAlert Storage Symbol Red
MDL Number MFCD00009171
Residue after Evaporation 0.001% max.
Identification Pass Test
Packaging NOWPak™ I
Fluorescence Background Pass Test
Water 0.02% max.
Synonym Acetic Acid Ethyl Ester
Health Hazard 3 Emergency Overview
Highly flammable. Vapors may cause drowsiness and dizziness. Repeated exposure may cause skin dryness or cracking. May cause central nervous system effects. May cause skin and respiratory tract irritation. May be harmful by inhalation, in contact with skin and if swallowed. Use only under a chemical fume hood. Use explosion-proof equipment. Wear personal protective equipment. Do not breathe vapors or spray mist. Do not get in eyes, on skin, or on clothing. Keep away from open flames, hot surfaces and sources of ignition. Keep away from open flames, hot surfaces and sources of ignition. Use only non-sparking tools.

NFPA

Health:1
Flammability:3
Instability:0
CAS 141-78-6
Linear Formula CH3COOC2H5
Molecular Formula C4H8O2
Chemical Name or Material Ethyl Acetate

Specifications

Chemical Name or Material Ethyl Acetate
CAS 141-78-6
Purity Grade HPLC Grade
Purity Grade Notes Mobile phase for HPLC applications
Assay ≥99.9%
Assay Percent Range Assay: 99.9% min by GC
Molecular Formula C4H8O2
Linear Formula CH3COOC2H5
Formula Weight 88.11
MDL Number MFCD00009171
Synonym Acetic Acid Ethyl Ester
Accessories
 

Emergency Overview
Highly flammable. Vapors may cause drowsiness and dizziness. Repeated exposure may cause skin dryness or cracking. May cause central nervous system effects. May cause skin and respiratory tract irritation. May be harmful by inhalation, in contact with skin and if swallowed. Use only under a chemical fume hood. Use explosion-proof equipment. Wear personal protective equipment. Do not breathe vapors or spray mist. Do not get in eyes, on skin, or on clothing. Keep away from open flames, hot surfaces and sources of ignition. Keep away from open flames, hot surfaces and sources of ignition. Use only non-sparking tools.

NFPA

Health:1
Flammability:3
Instability:0

DANGER!

 

SDS & Safety Documents