missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl Acetate, Exceeds A.C.S. Specifications, SpectroSolv (TM), 99.5%, Spectrum™
SP603, 141-78-6, C4H8O2
$1,148.60 - $4,155.80
Chemical Identifiers
CAS | 141-78-6 |
---|---|
Molecular Formula | C4H8O2 |
Molecular Weight (g/mol) | 88.11 |
InChI Key | XEKOWRVHYACXOJ-UHFFFAOYSA-N |
IUPAC Name | ethyl acetate |
SMILES | CCOC(C)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18602494
|
Spectrum Chemical Mfg Cor
SP603500MLCS6 |
6 x 500 mL | Amber Glass Bottle |
Case of 6 Each for $1,626.17
|
|
18602495
|
Spectrum Chemical Mfg Cor
SP6034LT |
4 L | Amber Glass Bottle |
Each for $1,148.60
|
|
18602496
|
Spectrum Chemical Mfg Cor
SP6034LTCS4 |
4 x 4 L | Amber Glass Bottle |
Case of 4 Each for $4,155.80
|
|
Description
Spectrum™ Ethyl Acetate, Exceeds A.C.S. Specifications, SpectroSolv (TM) is a colorless liquid that smells like pear drops. Abbreviated as EA, it can be found in glues, nail polish removers, cigarettes and other products. Primarily used as a diluent and solvent thanks in part to its low cost and toxicity it is also used in Laboratory settings for column chromatography and extractions. This SpectroSolv grade product is suitable for ultraviolet spectrophotometry usage and exceeds the specification of an ACS grade quality reagent. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.Chemical Identifiers
141-78-6 | |
88.11 | |
ethyl acetate |
C4H8O2 | |
XEKOWRVHYACXOJ-UHFFFAOYSA-N | |
CCOC(C)=O |
Specifications
141-78-6 | |
C4H8O2 | |
XEKOWRVHYACXOJ-UHFFFAOYSA-N | |
ethyl acetate | |
99.5% | |
0.003% | |
0.2% |
100% | |
UN1173 | |
CCOC(C)=O | |
88.11 | |
Amber Glass Bottle | |
0.0009 meq/g | |
6 x 500 mL |