missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl Acetate, Exceeds A.C.S. Specifications, HPLC Grade, 99.8%, Spectrum™
$261.89 - $6,892.00
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18610395
|
Spectrum Chemical Mfg Cor
HP6021LT |
1 L | Amber Glass Bottle |
Each for $261.89
|
|
18610396
|
Spectrum Chemical Mfg Cor
HP6024LT |
4 L | Amber Glass Bottle |
Each for $470.89
|
|
18610397
|
Spectrum Chemical Mfg Cor
HP602200LTBL |
200 L | Drum |
Each for $6,892.00
|
|
Chemical Identifiers
141-78-6 |
Specifications
141-78-6 | |
0.998 | |
Amber Glass Bottle | |
0.0009 meq/g | |
1 L |
1 | |
High Purity Solvent | |
3 ppm | |
0.0005 |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.