Please login to your online account to display your discounted pricing

Ethyl Acetate Ethanol Solution Optima™, 3:1, Fisher Chemical


Suitable for flash and normal phase chromatography

Quantity 1L

Manufacturer: fisher scientific company  E1511

 View more versions of this product

Catalog No. E1511

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Purity Grade OPTIMA™
Synonym EA/EtOH
Residue after Evaporation 10 ppm Maximum
Water 100 ppm Maximum
Color Colorless
Quantity 1L
CAS 141-78-6
Absorbance 0.01 AU Maximum(325 nm)
Storage Note 1 Flammable
Ethanol 24-26%
Chemical Name or Material Ethyl Acetate Ethanol Solution
Packaging Amber Glass
Physical Form Liquid
CAS 64-17-5

Specifications

Chemical Name or Material Ethyl Acetate Ethanol Solution
CAS 141-78-6
CAS 64-17-5
Purity Grade OPTIMA™
Synonym EA/EtOH

  • A green alternative to chlorinated solvents such as dichloromethane (DCM)
  • Suitable for use for flash and normal phase chromatography

Accessories

Flammable

SDS & Safety Documents