Please login to your online account to display your discounted pricing

Ethyl Acetate Ethanol Solution Optima™, 3:1, Fisher Chemical

Suitable for flash and normal phase chromatography

$499.92 - $1,112.33

Chemical Identifiers

Chemical Name or Material Ethyl Acetate Ethanol Solution
CAS 141-78-6
CAS 64-17-5
Purity Grade OPTIMA™
Synonym EA/EtOH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

E1511

  SDS
fisher scientific company
E1511
1L Each for $499.92
Case of 6 Each for $2,999.52

E1514

  SDS
fisher scientific company
E1514
4L Each for $1,112.33
Case of 4 Each for $4,449.32
Description & Specifications

Specifications

Chemical Name or Material Ethyl Acetate Ethanol Solution
Absorbance 0.01 AU Maximum(325 nm)
CAS 141-78-6
CAS 64-17-5
Storage Note 1 Flammable
Purity Grade OPTIMA™
Physical Form Liquid
Synonym EA/EtOH
Ethanol 24-26%
Packaging Amber Glass
Residue after Evaporation 10 ppm Maximum
Water 100 ppm Maximum
Color Colorless
Quantity 1L

Each lot is performance tested to ensure low UV absorbance for interference free chromatography

Safety and Handling

Flammable