missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl Acetate (Certified ACS), Fisher Chemical

Meet or exceeds the latest ACS specifications. Actual lot of analysis on label

Manufacturer:  Thermo Scientific E145S4

 View more versions of this product

Catalog No. E145S-4




Specifications

Specifications

Ethyl Acetate
Vapor Pressure: 103mbar at 20°C
C4H8O2
1173
XEKOWRVHYACXOJ-UHFFFAOYSA-N
ethyl acetate
8857
88.11
≥99.5%
0.902g/cm³
Aluminum Can
Pass Test
103mbar at 20°C
0.45 cP at 20°C
Colorless
4L
141-78-6
≥99.5 %
MFCD00009171
ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
CCOC(=O)C
88.106
CHEBI:27750
Liquid
Certified ACS Reagent
Pass Test
0.003% max.
0.0009mEq/g max.
0.1% max.
75°C
-83.5°C

Chemical Identifiers

141-78-6
88.106
XEKOWRVHYACXOJ-UHFFFAOYSA-N
8857
ethyl acetate
C4H8O2
MFCD00009171
ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
CHEBI:27750
CCOC(=O)C
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Highly flammable. Vapors may cause drowsiness and dizziness. Repeated exposure may cause skin dryness or cracking. May cause central nervous system effects. May cause skin and respiratory tract irritation. May be harmful by inhalation, in contact with skin and if swallowed. Use only under a chemical fume hood. Use explosion-proof equipment. Wear personal protective equipment. Do not breathe vapors or spray mist. Do not get in eyes, on skin, or on clothing. Keep away from open flames, hot surfaces and sources of ignition. Keep away from open flames, hot surfaces and sources of ignition. Use only non-sparking tools.

NFPA

Health:1
Flammability:3
Instability:0

DANGER!

DOTInformation : DOT Class 3, : Flammable Liquid

SDS Documents
Promotions

Promotions