Please login to your online account to display your discounted pricing

Ethyl Acetate (Certified ACS), Fisher Chemical


Meet or exceeds the latest ACS specifications. Actual lot of analysis on label

Quantity 1L
Packaging Glass Bottle

Manufacturer: fisher scientific company  E1451

 View more versions of this product

Catalog No. E145-1

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.5%
Boiling Point 75°C
Melting Point −83.5°C
Formula Weight 88.11
Quantity 1L
Health Hazard 1 DANGER!
DOT Information DOT Class 3, : Flammable Liquid
Physical Form Liquid
Vapor Pressure 103mbar at 20°C
Purity Grade Certified ACS Reagent Grade
Vapor Density 3.04g/mol
Viscosity 0.45 cP at 20°C
Color Colorless
Density 0.902g/cm³
UN Number 1173
Packaging Glass Bottle
Residue after Evaporation 0.003% max.
Assay Percent Range ≥99.5 %
Synonym Acetic Acid Ethyl Ester
Identification Pass Test
Additional Information Vapor Pressure: 103mbar at 20°C
Substances Darkened by H2SO4 Pass Test
Titratable Acid 0.0009mEq/g max.
Cleanup Symbol Solvent
Water 0.1% max.
MDL Number MFCD00009171
Chemical Name or Material Ethyl Acetate
Molecular Formula C4H8O2
CAS 141-78-6
ChemAlert Storage Symbol Red
Health Hazard 2 DANGER!
Health Hazard 3 Emergency Overview
Highly flammable. Vapors may cause drowsiness and dizziness. Repeated exposure may cause skin dryness or cracking. May cause central nervous system effects. May cause skin and respiratory tract irritation. May be harmful by inhalation, in contact with skin and if swallowed. Use only under a chemical fume hood. Use explosion-proof equipment. Wear personal protective equipment. Do not breathe vapors or spray mist. Do not get in eyes, on skin, or on clothing. Keep away from open flames, hot surfaces and sources of ignition. Keep away from open flames, hot surfaces and sources of ignition. Use only non-sparking tools.

NFPA

Health:1
Flammability:3
Instability:0

Specifications

Chemical Name or Material Ethyl Acetate
CAS 141-78-6
Purity Grade Certified ACS Reagent Grade
Assay ≥99.5%
Assay Percent Range ≥99.5 %
Molecular Formula C4H8O2
Formula Weight 88.11
MDL Number MFCD00009171
Synonym Acetic Acid Ethyl Ester
Accessories
 

DANGER!

Emergency Overview
Highly flammable. Vapors may cause drowsiness and dizziness. Repeated exposure may cause skin dryness or cracking. May cause central nervous system effects. May cause skin and respiratory tract irritation. May be harmful by inhalation, in contact with skin and if swallowed. Use only under a chemical fume hood. Use explosion-proof equipment. Wear personal protective equipment. Do not breathe vapors or spray mist. Do not get in eyes, on skin, or on clothing. Keep away from open flames, hot surfaces and sources of ignition. Keep away from open flames, hot surfaces and sources of ignition. Use only non-sparking tools.

NFPA

Health:1
Flammability:3
Instability:0

DANGER!

 

SDS & Safety Documents