Please login to your online account to display your discounted pricing

Ethyl Acetate, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$295.08 - $1,568.09

Chemical Identifiers

Chemical Name or Material Ethyl Acetate
CAS 141-78-6
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula CH3COOC2H5
Formula Weight 88.11
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002829

 
avantor performance
928022
Aluminum Bottle 1L Each for $295.08

02002834

 
avantor performance
928003
Narrow Mouth Amber Glass Bottle 4L Each for $618.19

02002835

 
avantor performance
928005
Aluminum Bottle 4L Each for $679.71

02002836

 
avantor performance
928033
Narrow Mouth PVC Coated Amber Bottle 4L Each for $1,568.09

02002839

 
avantor performance
928001
Narrow Mouth Amber Glass Bottle 500mL Case of 12 Each for $1,485.60
Description & Specifications

Specifications

Purity Grade Baker Analyzed™ ACS Reagent
Formula Weight 88.11
Chemical Name or Material Ethyl Acetate
CAS 141-78-6
Packaging Aluminum Bottle
Specific Gravity 1L = 0.90kg
Flash Point −4.00°C
Quantity 1L
Molecular Formula CH3COOC2H5