missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Specifications
Ethyl Acetate | |
141-78-6 | |
77.2°C | |
-4°C (-25°F) | |
Charactristic | |
Liquid | |
CH3COOCH2CH3 | |
ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester | |
XEKOWRVHYACXOJ-UHFFFAOYSA-N | |
ethyl acetate | |
8857 | |
88.11 | |
ACS Reagent |
-84°C | |
Colorless | |
20 L | |
425.85°C (798.5°F) | |
Metal Can | |
C4H8O2 | |
UN 1173 | |
Partially soluble in water | |
CCOC(=O)C | |
88.106 | |
CHEBI:27750 | |
≥99.5% |
Chemical Identifiers
141-78-6 | |
88.106 | |
ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester | |
CHEBI:27750 | |
CCOC(=O)C |
C4H8O2 | |
XEKOWRVHYACXOJ-UHFFFAOYSA-N | |
8857 | |
ethyl acetate |
Safety and Handling
DOTInformation : DOT Class: 3, Packaging Group: II