missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl acetate, ACS, 99.5+%

Manufacturer:  Alfa Aesar™ 31344M4

 View more versions of this product

Catalog No. AA31344M4



Specifications

Specifications

Ethyl Acetate
C4H8O2
MFCD00009171
506104
ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
CCOC(=O)C
88.106
CHEBI:27750
Liquid
ACS
Sweet
−3° (26°F)
Colorless
20L
141-78-6
CH3CO2CH2CH3
1173
14,3757
XEKOWRVHYACXOJ-UHFFFAOYSA-N
ethyl acetate
8857
88.11
≥99.5%
0.902g/mL
1.3720
77°C
-84°C

Chemical Identifiers

141-78-6
88.106
XEKOWRVHYACXOJ-UHFFFAOYSA-N
8857
ethyl acetate
C4H8O2
MFCD00009171
ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
CHEBI:27750
CCOC(=O)C
Safety and Handling

Safety and Handling

GHS H Statement
H225-H319-H336
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause drowsiness or dizziness.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHYL ACETATE

RTECSNumber : AH5425000

TSCA : TSCA

SDS Documents