missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl acetate, ACS, 99.5+%

Manufacturer:  Alfa Aesar™ 31344K7

 View more versions of this product

Catalog No. AA31344K7



Specifications

Specifications

Ethyl acetate
ACS Grade
C4H8O2
MFCD00009171
506104
XEKOWRVHYACXOJ-UHFFFAOYSA-N
ethyl acetate
8857
88.11
0.902g/mL
1.3720
77°C
-84°C
141-78-6
≥99.5%
CH3CO2CH2CH3
UN1173
14,3757
CCOC(=O)C
88.106
CHEBI:27750
Liquid
Sweet
−3° (26°F)
Colorless
4L

Chemical Identifiers

141-78-6
88.106
XEKOWRVHYACXOJ-UHFFFAOYSA-N
CHEBI:27750
C4H8O2
MFCD00009171
8857
Safety and Handling

Safety and Handling

GHS H Statement
H225-H319-H336
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause drowsiness or dizziness.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHYL ACETATE

RTECSNumber : AH5425000

TSCA : TSCA

SDS Documents