Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 9-fluorenylideneacetate, 98%

$227.81 - $227.81

Chemical Identifiers

Chemical Name or Material Ethyl 9-fluorenylideneacetate
CAS 27973-36-0
Molecular Formula C17H14O2
Formula Weight 250.3
MDL Number MFCD00798562
Synonym Dibenzofulvene-1-carboxylic acid ethyl ester; 9-Fluorenylideneacetic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1906218

  SDS
alfa aesar™
A1906218
50g Each for $227.81
Description & Specifications

Specifications

Formula Weight 250.3
Chemical Name or Material Ethyl 9-fluorenylideneacetate
Molecular Formula C17H14O2
CAS 27973-36-0
Assay 98%
MDL Number MFCD00798562
Synonym Dibenzofulvene-1-carboxylic acid ethyl ester; 9-Fluorenylideneacetic acid ethyl ester
Melting Point 75° to 77°C
Quantity 50g