missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 9-Bromononanoate, TCI America™

$288.75 - $988.50

Chemical Identifiers

CAS 28598-81-4
Molecular Formula C11H21BrO2
Molecular Weight (g/mol) 265.191
MDL Number MFCD06797065
InChI Key KXZBPLOPBKIUTC-UHFFFAOYSA-N
Synonym 9-Bromononanoic Acid Ethyl Ester
PubChem CID 560317
IUPAC Name ethyl 9-bromononanoate
SMILES CCOC(=O)CCCCCCCCBr
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B23241G View Documents TCI America
B23241G
1g Each for $288.75
 
B23245G View Documents TCI America
B23245G
5g Each for $988.50
 
Specifications

Chemical Identifiers

28598-81-4
265.191
KXZBPLOPBKIUTC-UHFFFAOYSA-N
560317
CCOC(=O)CCCCCCCCBr
C11H21BrO2
MFCD06797065
9-Bromononanoic Acid Ethyl Ester
ethyl 9-bromononanoate

Specifications

180°C
1g
C11H21BrO2
9-Bromononanoic Acid Ethyl Ester
CCOC(=O)CCCCCCCCBr
265.191
265.19
≥95.0% (GC)
Yellow
28598-81-4
MFCD06797065
KXZBPLOPBKIUTC-UHFFFAOYSA-N
ethyl 9-bromononanoate
560317
Clear Liquid at 20°C
Ethyl 9-Bromononanoate
Safety and Handling

Safety and Handling

TSCA : No