Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 7-bromoheptanoate, 97%

$60.58 - $60.58

Chemical Identifiers

Chemical Name or Material Ethyl 7-bromoheptanoate
CAS 29823-18-5
Molecular Formula Br(CH2)6CO2CH2CH3
Formula Weight 237.14
MDL Number MFCD00037103
Synonym 7-Bromoheptanoic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH28001AA

  SDS
alfa aesar™
H28001AA
5mL Each for $60.58
Description & Specifications

Specifications

Formula Weight 237.14
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Synonym 7-Bromoheptanoic acid ethyl ester
Health Hazard 1 Warning
Chemical Name or Material Ethyl 7-bromoheptanoate
Boiling Point 112°C (5mmHg)
Quantity 5mL
Molecular Formula Br(CH2)6CO2CH2CH3
MDL Number MFCD00037103
Flash Point >110°C (230°F)
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 97%
CAS 29823-18-5
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.