Learn More
Ethyl 7-bromoheptanoate, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H28001AA
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ethyl 7-bromoheptanoate | |
1.217 | |
>110°C (230°F) | |
MFCD00037103 | |
7-bromoheptanoic acid ethyl ester, ethyl-7-bromoheptanoate, heptanoic acid, 7-bromo-, ethyl ester, ethyl7-bromoheptanoate, br ch2 6cooet, ethyl 7-bromo-heptanoate, ethyl 7-bromanylheptanoate, acmc-209hb9, ethyl 7-bromoheptanoate, oobfndgmagsnka-uhfffaoysa | |
CCOC(=O)CCCCCCBr | |
237.14 | |
237.14 |
29823-18-5 | |
112°C (5 mmHg) | |
C9H17BrO2 | |
5 mL | |
OOBFNDGMAGSNKA-UHFFFAOYSA-N | |
ethyl 7-bromoheptanoate | |
122490 | |
97% |
Chemical Identifiers
29823-18-5 | |
237.14 | |
OOBFNDGMAGSNKA-UHFFFAOYSA-N | |
122490 | |
CCOC(=O)CCCCCCBr |
C9H17BrO2 | |
MFCD00037103 | |
7-bromoheptanoic acid ethyl ester, ethyl-7-bromoheptanoate, heptanoic acid, 7-bromo-, ethyl ester, ethyl7-bromoheptanoate, br ch2 6cooet, ethyl 7-bromo-heptanoate, ethyl 7-bromanylheptanoate, acmc-209hb9, ethyl 7-bromoheptanoate, oobfndgmagsnka-uhfffaoysa | |
ethyl 7-bromoheptanoate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P362-P501c
H315-H317-H319-H335
EINECSNumber : 249-883-6
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only