missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl 6-nitroindole-2-carboxylate, 97%, Thermo Scientific Chemicals
$625.58 - $625.58
Chemical Identifiers
CAS | 16792-45-3 |
---|---|
Molecular Formula | C11H10N2O4 |
Molecular Weight (g/mol) | 234.21 |
MDL Number | MFCD04973967 |
InChI Key | SOMZLSPJSSDTAP-UHFFFAOYSA-N |
Synonym | ethyl 6-nitroindole-2-carboxylate, 6-nitro-1h-indole-2-carboxylic acid ethyl ester, 6-nitro-2-carbethoxyindole, 6-nitro-2-carboethoxyindole, ethyl 6-nitroindol-2-carboxylate, 1h-indole-2-carboxylicacid, 6-nitro-, ethyl ester, 1h-indole-2-carboxylic acid, 6-nitro-, ethyl ester |
PubChem CID | 616444 |
IUPAC Name | ethyl 6-nitro-1H-indole-2-carboxylate |
SMILES | CCOC(=O)C1=CC2=CC=C(C=C2N1)[N+]([O-])=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6356903
|
Thermo Scientific Chemicals
H6356903 |
1 g |
Each for $625.58
|
|
|||||
Description
Ethyl 6-nitroindole-2-carboxylate used as a intermediate in pharmaceutical and chemical synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthyl 6-nitroindole-2-carboxylate used as a intermediate in pharmaceutical and chemical synthesis.
Solubility
Slightly soluble in water.
Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
Chemical Identifiers
16792-45-3 | |
234.21 | |
SOMZLSPJSSDTAP-UHFFFAOYSA-N | |
616444 | |
CCOC(=O)C1=CC2=CC=C(C=C2N1)[N+]([O-])=O |
C11H10N2O4 | |
MFCD04973967 | |
ethyl 6-nitroindole-2-carboxylate, 6-nitro-1h-indole-2-carboxylic acid ethyl ester, 6-nitro-2-carbethoxyindole, 6-nitro-2-carboethoxyindole, ethyl 6-nitroindol-2-carboxylate, 1h-indole-2-carboxylicacid, 6-nitro-, ethyl ester, 1h-indole-2-carboxylic acid, 6-nitro-, ethyl ester | |
ethyl 6-nitro-1H-indole-2-carboxylate |
Specifications
16792-45-3 | |
C11H10N2O4 | |
1 g | |
Slightly soluble in water. | |
CCOC(=O)C1=CC2=CC=C(C=C2N1)[N+]([O-])=O | |
234.21 | |
234.21 | |
Solid |
Yellow | |
MFCD04973967 | |
ethyl 6-nitroindole-2-carboxylate, 6-nitro-1h-indole-2-carboxylic acid ethyl ester, 6-nitro-2-carbethoxyindole, 6-nitro-2-carboethoxyindole, ethyl 6-nitroindol-2-carboxylate, 1h-indole-2-carboxylicacid, 6-nitro-, ethyl ester, 1h-indole-2-carboxylic acid, 6-nitro-, ethyl ester | |
SOMZLSPJSSDTAP-UHFFFAOYSA-N | |
ethyl 6-nitro-1H-indole-2-carboxylate | |
616444 | |
97% | |
Ethyl 6-nitroindole-2-carboxylate |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only