missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 6-nitroindole-2-carboxylate, 97%

Manufacturer:  Alfa Aesar™ H63569MD

 View more versions of this product

Catalog No. AAH63569MD



Description

Description

Applications
Ethyl 6-nitroindole-2-carboxylate used as a intermediate in pharmaceutical and chemical synthesis.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
Specifications

Specifications

Ethyl 6-nitroindole-2-carboxylate
250mg
C11H10N2O4
ethyl 6-nitroindole-2-carboxylate, 6-nitro-1h-indole-2-carboxylic acid ethyl ester, 6-nitro-2-carbethoxyindole, 6-nitro-2-carboethoxyindole, ethyl 6-nitroindol-2-carboxylate, 6-nitro-1h-indole-2-carboxylic acid ethyl ester, 1h-indole-2-carboxylicacid, 6-nitro-, ethyl ester, 1h-indole-2-carboxylic acid, 6-nitro-, ethyl ester
CCOC(=O)C1=CC2=C(N1)C=C(C=C2)[N+](=O)[O-]
234.211
234.21
97%
Yellow
16792-45-3
MFCD04973967
SOMZLSPJSSDTAP-UHFFFAOYSA-N
ethyl 6-nitro-1H-indole-2-carboxylate
616444
Solid

Chemical Identifiers

16792-45-3
234.211
SOMZLSPJSSDTAP-UHFFFAOYSA-N
616444
CCOC(=O)C1=CC2=C(N1)C=C(C=C2)[N+](=O)[O-]
C11H10N2O4
MFCD04973967
ethyl 6-nitroindole-2-carboxylate, 6-nitro-1h-indole-2-carboxylic acid ethyl ester, 6-nitro-2-carbethoxyindole, 6-nitro-2-carboethoxyindole, ethyl 6-nitroindol-2-carboxylate, 6-nitro-1h-indole-2-carboxylic acid ethyl ester, 1h-indole-2-carboxylicacid, 6-nitro-, ethyl ester, 1h-indole-2-carboxylic acid, 6-nitro-, ethyl ester
ethyl 6-nitro-1H-indole-2-carboxylate
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C

SDS Documents