Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 6-(chloroformyl)hexanoate, 95%

$263.69 - $1,155.69

Chemical Identifiers

Chemical Name or Material Ethyl 6-(chloroformyl)hexanoate
CAS 14794-32-2
Molecular Formula C9H15ClO3
Linear Formula ClCO-CH2CH2CH2CH2CH2CO2CH2CH3
Formula Weight 206.67
MDL Number MFCD00671507
Synonym 6-(Chloroformyl)hexanoic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5071406

  SDS
alfa aesar™
H5071406
5g Each for $1,155.69

AAH5071403

  SDS
alfa aesar™
H5071403
1g Each for $263.69
Description & Specifications

Specifications

Formula Weight 206.67
Chemical Name or Material Ethyl 6-(chloroformyl)hexanoate
Assay 95%
Molecular Formula C9H15ClO3
Linear Formula ClCO-CH2CH2CH2CH2CH2CO2CH2CH3
MDL Number MFCD00671507
CAS 14794-32-2
Synonym 6-(Chloroformyl)hexanoic acid ethyl ester
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g

Applications
Ethyl 6-(chloroformyl)hexanoate is used in agrochemical, pharmaceutical and dyestuff field.

Notes
Store in a cool, dry condition in a well sealed container. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.