Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 6-bromoimidazo[1,2-a]pyridine-2-acetate, 98%

Chemical Identifiers

Chemical Name or Material Ethyl 6-bromoimidazo[1,2-a]pyridine-2-acetate
CAS 59128-04-0
Formula Weight 283.13
MDL Number MFCD16172180
Beilstein 791271
Synonym 6-Bromoimidazo[1,2-a]pyridine-2-acetic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6443003

 
alfa aesar™
H6443003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6443006

 
alfa aesar™
H6443006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH64430MD

 
alfa aesar™
H64430MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 55° to 56°C
Formula Weight 283.13
Chemical Name or Material Ethyl 6-bromoimidazo[1,2-a]pyridine-2-acetate
Physical Form Solid
Assay 98%
Color Gray
Quantity 1g
MDL Number MFCD16172180
CAS 59128-04-0
Beilstein 791271
Synonym 6-Bromoimidazo[1,2-a]pyridine-2-acetic acid ethyl ester
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.