missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 6-Bromocoumarin-3-carboxylate, TCI America™

Manufacturer:  TCI America E073325G

 View more versions of this product

Catalog No. E073325G



Specifications

Specifications

Ethyl 6-Bromocoumarin-3-carboxylate
169°C
2199-90-8
MFCD00124584
RXGDNKCWEKUYQA-UHFFFAOYSA-N
ethyl 6-bromo-2-oxochromene-3-carboxylate
691747
Crystal-Powder at 20°C
White-Yellow
25g
C12H9BrO4
6-Bromocoumarin-3-carboxylic Acid Ethyl Ester
CCOC(=O)C1=CC2=C(C=CC(=C2)Br)OC1=O
297.104
297.10
≥98.0% (GC)

Chemical Identifiers

2199-90-8
297.104
RXGDNKCWEKUYQA-UHFFFAOYSA-N
691747
CCOC(=O)C1=CC2=C(C=CC(=C2)Br)OC1=O
C12H9BrO4
MFCD00124584
6-Bromocoumarin-3-carboxylic Acid Ethyl Ester
ethyl 6-bromo-2-oxochromene-3-carboxylate
Safety and Handling

Safety and Handling

TSCA : No

Documents