Please login to your online account to display your discounted pricing

Ethyl 6-bromo-2,7-dihydro-4-methyl-2,7-dioxo-3h-dibenzo[f,ij]Isoquinolinecarboxylate, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Ethyl 6-bromo-2,7-dihydro-4-methyl-2,7-dioxo-3H-dibenzo[f,ij] isoquinolinecarboxylate
Assay Percent Range 98%
Molecular Formula C20H14BrNO4
Formula Weight 412.23
MDL Number MFCD02728506
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC374020010

  SDS
thermo fisher scientific
AC374020010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC374020050

  SDS
thermo fisher scientific
AC374020050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 412.23
Physical Form Crystalline Powder
Quantity 1g
Color Brown
Elemental Analysis 97.5% min.
CAS 51418-86-1
Molecular Formula C20H14BrNO4
MDL Number MFCD02728506
Chemical Name or Material Ethyl 6-bromo-2,7-dihydro-4-methyl-2,7-dioxo-3H-dibenzo[f,ij] isoquinolinecarboxylate
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 98%
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.