missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 6-Aminopyridine-2-carboxylate, TCI America™

Manufacturer:  TCI America E12131G

 View more versions of this product

Catalog No. E12131G



Specifications

Specifications

Ethyl 6-Aminopyridine-2-carboxylate
72°C
69142-64-9
MFCD01009842
DTYDENRYZSEGNL-UHFFFAOYSA-N
ethyl 6-aminopyridine-2-carboxylate
11959065
Crystal-Powder at 20°C
Yellow
1g
C8H10N2O2
6-Aminopyridine-2-carboxylic Acid Ethyl Ester, Ethyl 6-Aminopicolinate, 6-Aminopicolinic Acid Ethyl Ester
CCOC(=O)C1=NC(=CC=C1)N
166.18
166.18
≥98.0% (GC,T)

Chemical Identifiers

69142-64-9
166.18
DTYDENRYZSEGNL-UHFFFAOYSA-N
11959065
CCOC(=O)C1=NC(=CC=C1)N
C8H10N2O2
MFCD01009842
6-Aminopyridine-2-carboxylic Acid Ethyl Ester, Ethyl 6-Aminopicolinate, 6-Aminopicolinic Acid Ethyl Ester
ethyl 6-aminopyridine-2-carboxylate
Safety and Handling

Safety and Handling

TSCA : No

Documents