Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl 6-amino-5-methoxyindole-2-carboxylate, 95%

$163.16 - $657.27

Chemical Identifiers

Chemical Name or Material Ethyl 6-amino-5-methoxyindole-2-carboxylate
CAS 107575-60-0
Molecular Formula C12H14N2O3
Formula Weight 234.25
MDL Number MFCD08064224
Synonym 6-Amino-5-methoxyindole-2-carboxylic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2554903

 
alfa aesar™
B2554903
1g Each for $163.16

AAB2554906

 
alfa aesar™
B2554906
5g Each for $657.27
Description & Specifications

Specifications

Formula Weight 234.25
Chemical Name or Material Ethyl 6-amino-5-methoxyindole-2-carboxylate
Assay 95%
Molecular Formula C12H14N2O3
MDL Number MFCD08064224
CAS 107575-60-0
Synonym 6-Amino-5-methoxyindole-2-carboxylic acid ethyl ester
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 167° to 169°C
Quantity 1g

Applications
Ethyl 6-amino-5-methoxyindole-2-carboxylate is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in dichloromethane, ethyl acetate and methanol.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.