missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 6-[4-(Diphenylamino)phenyl]coumarin-3-carboxylate, TCI America™

Manufacturer:  TCI America E0783200MG

 View more versions of this product

Catalog No. E0783200MG



Specifications

Specifications

Ethyl 6-[4-(Diphenylamino)phenyl]coumarin-3-carboxylate
200mg
C30H23NO4
PQYVEQOPURSPHF-UHFFFAOYSA-N
ethyl 2-oxo-6-[4-(N-phenylanilino)phenyl]chromene-3-carboxylate
44629967
Crystal-Powder at 20°C
Undesignated
1056693-13-0
6-[4-(Diphenylamino)phenyl]coumarin-3-carboxylic Acid Ethyl Ester, 3-Ethoxycarbonyl-6-[4-(diphenylamino)phenyl]coumarin
CCOC(=O)C1=CC2=C(C=CC(=C2)C3=CC=C(C=C3)N(C4=CC=CC=C4)C5=CC=CC=C5)OC1=O
461.517
461.52
≥98.0% (HPLC)

Chemical Identifiers

1056693-13-0
461.517
6-[4-(Diphenylamino)phenyl]coumarin-3-carboxylic Acid Ethyl Ester, 3-Ethoxycarbonyl-6-[4-(diphenylamino)phenyl]coumarin
ethyl 2-oxo-6-[4-(N-phenylanilino)phenyl]chromene-3-carboxylate
C30H23NO4
PQYVEQOPURSPHF-UHFFFAOYSA-N
44629967
CCOC(=O)C1=CC2=C(C=CC(=C2)C3=CC=C(C=C3)N(C4=CC=CC=C4)C5=CC=CC=C5)OC1=O
Safety and Handling

Safety and Handling

TSCA : No

Documents