missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl 5H-Octafluorovalerate, TCI America™

Manufacturer:  TCI America E102225G

 View more versions of this product

Catalog No. E102225G



Specifications

Specifications

Ethyl 5H-Octafluorovalerate
Colorless
2795-50-8
MFCD00153153
5H-Octafluorovaleric Acid Ethyl Ester
CCOC(=O)C(C(C(C(F)F)(F)F)(F)F)(F)F
274.11
274.11
≥98.0% (GC)
180°C
25g
C7H6F8O2
3272
HXWMNJVBQLBDGW-UHFFFAOYSA-N
ethyl 2,2,3,3,4,4,5,5-octafluoropentanoate
2737223
Clear Liquid at 20°C

Chemical Identifiers

2795-50-8
274.11
HXWMNJVBQLBDGW-UHFFFAOYSA-N
2737223
CCOC(=O)C(C(C(C(F)F)(F)F)(F)F)(F)F
C7H6F8O2
MFCD00153153
5H-Octafluorovaleric Acid Ethyl Ester
ethyl 2,2,3,3,4,4,5,5-octafluoropentanoate
Safety and Handling

Safety and Handling

TSCA : No

Documents